missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- LC-Gradient with Diode Array Detection (AU), 0.1% (v/v) aqueous solution:
- at 220 nm max. 0.002
- at 254 nm max. 0.002
- LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), 0.1% (v/v) aqueous solution:
- Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ppb
- Ultraviolet Absorbance, 0.1% (v/v) aqueous solution (1.00-path vs water):
- at 240 nm max. 0.1
- at 260 nm max. 0.01
Specifications
Specifications
| Assay Percent Range | min. 99% |
| UN Number | 1779 |
| Quantity | 10 mL |
| Flash Point | 69°C |
| Packaging | Glass bottle |
| Residue after Evaporation | max. 20 ppm |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?