missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99%, for molecular biology, DNAse, RNAse and Protease free

Product Code. 10114763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10114763 2.5 L Glass Bottle 2.5 L
10602882 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10114763 Supplier Thermo Scientific Acros Supplier No. 327230025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-12-7 | CH3NO | 45.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For molecular biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

pH 7 to 9 (1M in 0.1M KCl)
Melting Point 2.0°C to 3.0°C
Absorbance (0.5 M in water), (0.5 M in water) at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Density 1.1330g/mL
Boiling Point 210.0°C
Quantity 2.5 L
Flash Point 175°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4460 to 1.4480
Assay Percent Range 99%
Specific Gravity 1.133
Linear Formula HCONH2
Viscosity 3.75 mPa.s (20°C)
Beilstein 02, 26
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Formula Weight 45.04
Percent Purity 99%
Grade Molecular Biology
Chemical Name or Material Formamide, DNase, RNase and Protease Free
Show More Show Less
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Formamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.