Learn More
Karl Hecht™ Assistent™ Mechanical Flow Monitor
Product Code. 10145961
Change view
Click to view available options
:
Inner Diameter: 6 to 11mm
Product Code.
10145961 Inner Diameter: 6 to 11mm
1 options
Product Code. 10145961 Supplier Karl Hecht™ Supplier No. 43178010
Only null left
Add to basket
Add to basket
For control of the water flow in thermostatic water baths. Karl Hecht™ Assistent™ Mechanical Flow Monitor is manufactured from translucent plastic (SAN).
  • For tubing of 6-11mm inside diameters
  • Brightly colored paddle wheel spins freely when flow occurs
Specifications
Product Type Flow Switch