missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Flexible Shaft Laboral Retort Clamp for Burettes

Product Code. 10064900
Change view
Click to view available options
Diameter (Metric):
0 to 30 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10064900 0 to 30 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10064900 Supplier Bochem™ Supplier No. B08303033

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Quantity 1 Ea.
Type Burette Clamp
Diameter (Metric) 0 to 30 mm
Length (Metric) 220 mm
Diameter (Metric) Inside 0 to 30 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.