Learn More
BOLA Flat Flange Joining Pieces
Description
- Made of aluminium
- Connection between reaction vessel and lid.
- Locked by zinc-plated steel screws.
Specifications
Specifications
| Standard Taper Joint | 170/146 |
| Joint Type | 170/ 146 |
| Autoclavable | Autoclavable |