missing translation for 'onlineSavingsMsg'
Learn More

FL-SMRT ID1 Peptide, 100μM

Product Code. 10093143
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10093143 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10093143 Supplier Invitrogen Supplier No. PV4620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

FL-SMRT ID1 PEPTIDE, 100UM

FL-SMRT ID1 PEPTIDE
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 100 μM Fluorescein-SMRT ID1 Peptide in 99.9% DMSO, 0.1% TFA. Store at -20°C and protect from light.
Label Type Classic Dyes
Quantity 100 μL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.