missing translation for 'onlineSavingsMsg'
Learn More

Kartell SpA Fixopet FX 250

Product Code. 10030772
Change view
Click to view available options
Volume (Metric):
250 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10030772 250 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030772 Supplier Kartell SpA Supplier No. 0131010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fixed volume autoclavable micropipettes

High precision Fixed volume pipette. Equipped with adjustable tip ejector in order to eliminate any risk of contamination. The structural feature considerably reduces the force necessary for pipetting. Ideal for clinical and educational uses. the integrated recalibration system ensures quick and precise recalibrations

Robust structure of the body made of PVDF ensures high mechanical and chemical resistances and permit exposure to UV rays

In accordance with EN IS 8655

Specifications

Autoclavable Autoclavable
Color Red
Disposable Reusable
For Use With Micropipette Tip
Material Plastic
Model Fixopet
Number of Channels 1
Sterility Non-sterile
Type Fixed Volume Micropipette
Volume (Metric) 250 μL
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.