Learn More
Value Plastics Fitting Manifold with 200 Series Barbs, Polypropylene
Product Code. 30053130 Shop All Cole-Parmer Products
Change view
Click to view available options
:
25 Hose Barb Fitting Manifold 3-Ports PP 1/8 in.
Product Code.
30053130 25 Hose Barb Fitting Manifold 3-Ports PP 1/8 in.
1 options
Product Code. 30053130 Supplier Value Plastics Supplier No. undefined
Only null left
Add to basket
Add to basket
Ensure efficient fluid distribution while minimizing pressure drops
Constructed of ADCF polypropylene. Utilize 200 Series barbs in high pressure applications. Manufactured to strict quality standards under an ISO 9001:2000 quality control
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Packaging Quantity 25/Pk.
Outlet Port Hose Barb Union Manifold
Sterilization Method 121°C
Shape Manifold