missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Firefly luciferase Monoclonal Antibody (CS 17)
GREENER_CHOICE

Product Code. 10027812
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10027812 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10027812 Supplier Invitrogen™ Supplier No. 356700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

Reactivity was confirmed via Western Blot analysis on the following samples containing added purified luciferase protein: mouse heart, liver, kidney, and cortex tissues. There was no cross reactivity to mouse heart, NIH3T3, Jurkat, and human fibroblasts. In IHC (frozen tissue) and Immunofluorescence applications, this antibody is able to detect tissue specific expression of Luciferase protein in Drosophila embryos using the well-characterized Gal4-UAS system with two different Gal4 drivers.

Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Firefly luciferase
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone CS 17
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase
Host Species Mouse
Immunogen Luciferase protein from Photinus pyralis.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Luciferase
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.