Learn More
Value Plastics Finger Snap Luer Lock Ring, Polycarbonate
Shop All Cole-Parmer Products
Click to view available options
:
25 Finger Snap Male Luer Lock Ring PC
Description
Luer fittings allow for a simple yet secure twist-lock connection. Constructed of FDA compliant clear polycarbonate. Prevent leaks with lock ring mechanism
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS;FDA |
| Packaging Quantity | 25/Pk. |
| Inlet | Finger Snap Luer Lock Ring |