Learn More
Thermo Scientific™ NoLimits 600 bp DNA Fragment
GREENER_CHOICE
Product Code. 10170780 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
10170780 10 μg 10µg
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10170780 Supplier Thermo Scientific™ Supplier No. SM1461
Edge
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
NoLimits is a collection of 36 chromatography-purified individual DNA fragments ranging from 10 to 20,000 bp for applications where a custom DNA size standard is required.
Thermo Scientific NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology. Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
TRUSTED_SUSTAINABILITY
Specifications
Gel Type Agarose
Ready to Load No
Size Range 10 to 20000 bp
Gel Compatibility Agarose Gels
Quantity 10 μg
Product Line NoLimits
Product Type DNA Fragment