missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aminoallyl-UTP Solution (50 mM)

Product Code. 10121080 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121080 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10121080 Supplier Thermo Scientific™ Supplier No. R1091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Thermo Scientific Aminoallyl-UTP (5-[3-aminoallyl]-2'-uridine-5'-triphosphate) is supplied as 50 mM aqueous solution titrated to pH 7.0 with NaOH.

Thermo Scientific Aminoallyl-UTP (5-[3-aminoallyl]-2'-uridine-5'-triphosphate) is supplied as 50 mM aqueous solution titrated to pH 7.0 with NaOH.

Aminoallyl-UTP can be enzymatically incorporated into RNA with T7 (Cat. No. EP0111), T3 (EP0101) and SP6 RNA polymerases (EP0131). The resulting amine-containing RNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin, or hapten.

Applications
• Indirect non-radioactive enzymatic labeling of RNA during in vitro transcription

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Aminoallyl-UTP Solution, 50 mM (50 μL)

Store at –20°C.
Concentration 50 mM
For Use With (Application) Non-Radioactive Labeling
Labeling Method Direct Labeling
Label or Dye Aminoallyl
Product Type Aminoallyl-UTP Solution
Quantity 50 μL
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.