missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ FEP Coated Silicone O-Rings for Flat Flanges

Product Code. 10114541
Change view
Click to view available options
For Use With (Equipment):
Flat flanges with groove
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10114541 Flat flanges with groove Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10114541 Supplier Bohlender™ Supplier No. H96918
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For sealing a flat flange with groove.

  • Providing temperature resistance from −60° to +205°C
  • Very good chemical resistance
  • FDA conformity
  • Silicone core with seamless FEP coating
  • Manufactured according to DIN 12214; 1996-12
  • Flexible
  • For flat flange NS 60
  • Dimensions: 75 x 4 mm

Specifications

For Use With (Application) Sealing flat flanges
For Use With (Equipment) Flat flanges with groove
Product Type O-Ring
Material FEP-coated silicone
Color Red
Description For flat flange NW 60
Size 75 x 4 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.