missing translation for 'onlineSavingsMsg'
Learn More

Fenpyroximate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30309699
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309699 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309699 Supplier Dr. Ehrenstorfer Supplier No. DREV13545300AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenpyroximate (E/Z Mix)
CAS 111812-58-9
Analyte or Component Names Fenpyroximate (E/Z Mix)
Shipping Condition Room Temperature
Molecular Formula C24 H27 N3 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzoic acid, 4-[[[[(1,3-dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]-, 1,1-dimethylethyl ester, Fenpyroximate
InChI Formula InChI=1S/C24H27N3O4/c1-17-21(22(27(5)26-17)30-20-9-7-6-8-10-20)15-25-29-16-18-11-13-19(14-12-18)23(28)31-24(2,3)4/h6-15H,16H2,1-5H3
SMILES Cc1nn(C)c(Oc2ccccc2)c1C=NOCc3ccc(cc3)C(=O)OC(C)(C)C
IUPAC Name tert-butyl 4-[[(1,3-dimethyl-5-phenoxypyrazol-4-yl)methylideneamino]oxymethyl]benzoate
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 421.49
Quantity 5 mL
Formula Weight 421.2002 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.