missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenoprofen Isopropyl Ester, Mikromol™
Description
Fenoprofen Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Fenoprofen Isopropyl Ester |
| CAS | 120824-55-7 |
| Analyte or Component Names | Fenoprofen Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H20 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Fenoprofen Isopropyl Ester, alpha-Methyl-3-phenoxybenzeneacetic acid 1-methylethyl ester, Isopropyl 2-(3-phenoxyphenyl)propionate |
| InChI Formula | InChI=1S/C18H20O3/c1-13(2)20-18(19)14(3)15-8-7-11-17(12-15)21-16-9-5-4-6-10-16/h4-14H,1-3H3 |
| SMILES | CC(C)OC(=O)C(C)c1cccc(Oc2ccccc2)c1 |
| IUPAC Name | propan-2-yl 2-(3-phenoxyphenyl)propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?