Learn More
Socorex™ Feed Tubing, Socorex™ 187 Syringe
Description
- Fits Socorex™ 187 self-refilling syringe.
- Long lasting.
- Medical quality silicone construction.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Silicone |
| For Use With | Socorex 187 Tube Feeding Syringe |
| Flow Capacity | 0.3 to 10 mL |
| Product Type | Feed Tubing |