Learn More
Testo™ Fast-action Immersion/Penetration Probe
Product Code. 10163053
Change view
Click to view available options
Length (Metric) Probe:
60 mm
Unit Size:
Each
Product Code. Length (Metric) Probe unitSize
10163053 60 mm Each
1 options
Product Code. 10163053 Supplier Testo™ Supplier No. 0602 2693
Only null left
Add to basket
Add to basket
Specifications
Includes Immersion/Penetration Probe, 5 dia. x 60mm L Probe Shaft, 1.5 dia. x 40mm L Probe Shaft Tip, 1.2m L Cable, –60 to 800°C Measuring Range, 3 sec. Response Time, For Temperature Measurements in Pastes and Semi-solid Media, in Liquids and in the Air.
Certifications/Compliance EN 60584-2
For Use With (Application) Temperature Measurements in Pastes and Semi-solid Media, in Liquids and in the Air
Response Time 3 sec.
Temperature Range (Metric) -60°C to +800°C
Length (Metric) Probe 60 mm