missing translation for 'onlineSavingsMsg'
Learn More

Fadraciclib, MedChemExpress

Product Code. 16494546
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494546 100 mg 100mg
16483298 10 mg 10mg
16479836 50 mg 50mg
16460739 1 mg 1mg
16406027 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494546 Supplier Medchem Express Supplier No. HY101212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fadraciclib (CYC065) is a second-generation, orally available ATP-competitive inhibitor of CDK2/CDK9 kinases with IC50s of 5 and 26 nM, respectively.

Fadraciclib (CYC065) is a second-generation, orally available ATP-competitive inhibitor of CDK2/CDK9 kinases with IC50s of 5 and 26 nM, respectively.

Specifications

Chemical Name or Material Fadraciclib
CAS 1070790-89-4
Color White
Purity Grade Notes Research
Molecular Formula C21H31N7O
Quantity 100 mg
Synonym CYC065
Solubility Information DMSO : ≥ 100 mg/mL (251.56 mM)
SMILES C[C@@H](O)[C@@H](NC1=NC(NCC2=C(C)C=C(C)N=C2)=C3N=CN(C(C)C)C3=N1)CC
Molecular Weight (g/mol) 397.52
Formula Weight 397.52
Percent Purity 99.56%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.