missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor XIII Recombinant Rabbit Monoclonal Antibody (K01_1K61)
Description
Factor XIII Recombinant Monoclonal Antibody for IHC (P), Western Blot
This gene encodes the coagulation factor XIII A subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon cleavage of the activation peptide by thrombin and in the presence of calcium ion, the plasma factor XIII dissociates its B subunits and yields the same active enzyme, factor XIIIa, as platelet factor XIII. This enzyme acts as a transglutaminase to catalyze the formation of gamma-glutamyl-epsilon-lysine crosslinking between fibrin molecules, thus stabilizing the fibrin clot. It also crosslinks alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin. Factor XIII deficiency is classified into two categories: type I deficiency, characterized by the lack of both the A and B subunits; and type II deficiency, characterized by the lack of the A subunit alone. These defects can result in a lifelong bleeding tendency, defective wound healing, and habitual abortion.
Specifications
Specifications
| Antigen | Factor XIII |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1K61 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | F13A1 |
| Gene Accession No. | P00488 |
| Gene Alias | bA525O21.1 (coagulation factor XIII, A1 polypeptide); coagulation factor XIII; coagulation factor XIII A chain; Coagulation factor XIII B chain; coagulation factor XIII, A polypeptide; coagulation factor XIII, A1 polypeptide; coagulation factor XIII, B polypeptide; coagulation factor XIIIa; F13; F13A; F13A1; F13B; Factor 13; factor viiia; factor XIIIa; Factor13; fibrin stabilizing factor, A subunit; fibrinoligase; fibrin-stabilizing factor B subunit; FSF, A subunit; FXIIIB; Protein-glutamine gamma-glutamyltransferase A chain; protein-glutamine gamma-glutamyltransferase B chain; TGase; transglutaminase A chain; Transglutaminase B chain; transglutaminase. plasma |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?