Learn More
Invitrogen™ F(ab')2-Goat anti-Human IgG Fc gamma Secondary Antibody
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31163 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31163 reacts with the heavy chains of human IgG, but not with the light chains to most human immunoglobulins or with non-immunoglobulin serum proteins. This antibody does not react against human IgM or IgA. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough product for a single day's use. Store remainder at 2-8 °C under sterile conditions. Country of Origin: USA.
Specifications
Specifications
| Antigen | Human IgG Fc gamma |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.