Learn More
Lenz Laborglasintrumente Extractors for Specific Heavy Solvents
Description
- Extractor head with insert filter (frit, porosity: 0), with PTFE valve stopcock
- Dimroth condenser with thread GL 14
- Round bottom flask
- DURAN™
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Porosity | 0 |
| Standard Taper Joint Side | NS 60/46 |