Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Heavy Solvents
Description
- For the extraction of liquids with specific heavy solvents
- With insert filter (porosity: 0)
- Made of DURAN™ tubing
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Fritte P0 |
| Standard Taper Joint | NS 45/40 |