missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SuperScript™ EXPRESS One-Step Superscript™ qRT-PCR Kit, with premixed ROX
Description
EXPRESS One-Step SuperScript™ qRT-PCR SuperMix Kits incorporate proven Invitrogen enzymes now specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for your probe-based qPCR experiments.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase - complete, rapid activation provides the best sensitivity and reproducibility
- The One-Step SuperScript™ III Reverse Transcriptase module has a unique formulation to ensure better yield and less PCR inhibition, specifically for one-step qRT-PCR
- Flexible formats with ROX premixed or separate - designed to work with default instrument protocols on a wide range of high-throughput fast instruments (e.g., AB 7500 Fast, AB 7900 HT Fast, AB StepOne, Roche LightCycler™ 480, Corbett Rotor-Gene™)
- The only commercially available one-step qRT-PCR kits containing heat-labile UDG - significantly reduced carryover contamination risk with excellent activity from room temperature to approximately 40°C. Due to the thermostability of SuperScript™ III, the cDNA synthesis step is run at 50°C, at which temperature the heat-labile UDG is totally and irreversibly inactivated
Easy to test and switch:
- EXPRESS kits work with manufacturer's default protocols on fast instruments and come in a convenient and stable SuperMix format for ultimate convenience
- The kits are offered with either premixed ROX or a separate tube of ROX to fit your instrument needs
Order Info
Guaranteed to be stable for 6 months when properly stored.
Specifications
Specifications
| Content And Storage | EXPRESS One-Step SuperScript™ qRT-PCR SuperMixes are composed of 2 modules: an EXPRESS RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and an EXPRESS qPCR SuperMix module with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed stable for 6 months when properly stored. |
| Detection Method | Primer-probe |
| Format | Tube |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Fast or Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode |
| Product Line | SuperScript |
| Product Type | qRT-PCR Kit |
| Quantity | 2,500 reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?