missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Exendin-3

Product Code. 16442188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442188 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442188 Supplier MedChemExpress Supplier No. HYP1543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Exendin-3 is a biologically active peptides isolated from venoms of the Gila monster lizards, Heloderma horridurn.

Specifications

CAS 130357-25-4
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 4202.57 g/mol
Product Type Natural Products
Quantity 5 mg
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.