missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylnoradrenaline Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Ethylnoradrenaline Hydrochloride |
| CAS | 3198-07-0 |
| Analyte or Component Names | Ethylnoradrenaline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H15 N O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,2-Benzenediol, 4-(2-amino-1-hydroxybutyl)-, hydrochloride (1:1), 1,2-Benzenediol, 4-(2-amino-1-hydroxybutyl)-, hydrochloride (9CI), Bronkephrine, Butanephrine hydrochloride, Ethylnorepinephrine Hydrochloride, Benzyl alcohol, alpha-(1-aminopropyl)-3,4-dihydroxy-, hydrochloride (7CI,8CI), Protocatechuyl alcohol, alpha-(1-aminopropyl)-, hydrochloride (6CI), alpha-(1-Aminopropyl)protocatechuyl alcohol hydrochloride, alpha-Ethylnorepinephrine hydrochloride |
| InChI Formula | InChI=1S/C10H15NO3.ClH/c1-2-7(11)10(14)6-3-4-8(12)9(13)5-6;/h3-5,7,10,12-14H,2,11H2,1H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCC(N)C(O)c1ccc(O)c(O)c1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?