missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic-d12 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | EDTA-D12 |
| CAS | 203806-08-0 |
| Molecular Formula | C10 D12 H4 N2 O8 |
| Quantity | 0.1 g |
| Synonym | Glycine-2,2-d2, N,N'-(1,2-ethanediyl-1,1,2,2-d4)bis[N-(carboxymethyl-d2)- (9 CI, ACI), N,N'-(1,2-Ethanediyl-1,1,2,2-d4)bis[N-(carboxymethyl-d2)glycine-2,2-d2] (ACI), Ethylenediamine-N,N,N',N'-tetraacetic Acid-d12, Ethylene-d4-diamine-N,N,N',N'-tetraacetic-2,2-d2 acid, EDTA-d12, Edetic acid-D12 |
| InChI Formula | InChI=1 S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)/i1D2,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])(N(C([2 H])([2 H])C(=O)O)C([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C(=O)O)C([2 H])([2 H])C(=O)O)C(=O)O |
| IUPAC Name | 2-[[2-[bis[carboxy(dideuterio)methyl]amino]-1,1,2,2-tetradeuterioethyl]-[carboxy(dideuterio)methyl]amino]-2,2-dideuterioacetic acid |
| Molecular Weight (g/mol) | 304.3166 |
| Formula Weight | 304.166 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction