missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Product Code. 10089770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10089770 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10089770 Supplier Thermo Scientific Chemicals Supplier No. 327211000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13235-36-4 | C10H20N2Na4O12 | 452.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 13235-36-4
Absorbance 0.06 max. (0.1 M in water) at 280nm, 1.5 max. (0.1 M in water) at 260nm
Color White
pH 10.7 to 11.7 (0.1M aq. soln.)
Flash Point >100°C
Assay Percent Range 99.4% min. (Complexometry)
Packaging Glass bottle
Molecular Formula C10H20N2Na4O12
MDL Number MFCD00150027
Quantity 100 g
Fieser 01,373; 05,296
Merck Index 14, 3517
Synonym tetrasodium ethylenediaminetetraacetate tetrahydrate, edetate sodium tetrahydrate, unii-l13nhd21x6, ethylenediaminetetraacetic acid tetrasodium salt tetrahydrate, acmc-209bp2, ksc910c4l, edta, tetrasodium tetrahydrate salt, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tetrasodium salt, tetrahydrate, ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, sodium salt, hydrate 1:4:4
Solubility Information (0.1 M in water) Clear colorless
InChI Key XFLNVMPCPRLYBE-UHFFFAOYSA-J
SMILES O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
IUPAC Name tetrasodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
Molecular Weight (g/mol) 452.23
PubChem CID 23287279
Formula Weight 452.23
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.