missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-(4-chlorophenyl)-2-formyl-3-furoate, 97%, Thermo Scientific™

Product Code. 10138754
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10138754 5 g 5g
10792323 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10138754 Supplier Thermo Scientific Maybridge Supplier No. SP00067DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175276-64-9
Molecular Formula C14H11ClO4
Molecular Weight (g/mol) 278.688
MDL Number MFCD00275550
InChI Key XFOQMTOCKSVCBO-UHFFFAOYSA-N
PubChem CID 2798516
IUPAC Name ethyl 5-(4-chlorophenyl)-2-formylfuran-3-carboxylate
SMILES CCOC(=O)C1=C(OC(=C1)C2=CC=C(C=C2)Cl)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Synonym ethyl 5-4-chlorophenyl-2-formyl-3-furoate, ethyl 5-4-chlorophenyl-2-formylfuran-3-carboxylate, ethyl5-4-chlorophenyl-2-formyl-3-furoate, 3-furancarboxylic acid,5-4-chlorophenyl-2-formyl-, ethyl ester, 3-furancarboxylic acid,5-4-chlorophenyl-2-formyl-,ethyl ester
Formula Weight 278.69
Percent Purity 97%
Chemical Name or Material ethyle5-(4-chlorophenyl)-2-formyl-3-furoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.