missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Pantothenate, Mikromol™

Product Code. 30050956
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050956 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050956 Supplier Mikromol Supplier No. MM1499.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl Pantothenate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Pantothenate
CAS 10527-68-1
Analyte or Component Names Ethyl Pantothenate
Shipping Condition Room Temperature
Molecular Formula C11 H21 N O5
Certifications/Compliance ISO 9001
Synonym Ethyl Pantothenate, N-[(2R)-2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl]-beta-alanine ethyl ester, Pantothenic acid, ethyl ester (7CI,8CI), beta-Alanine, N-(2,4-dihydroxy-3,3-dimethyl-1-oxobutyl)-, ethyl ester, (R)-, Ethyl D-pantothenate, Ethyl pantothenate
InChI Formula InChI=1S/C11H21NO5/c1-4-17-8(14)5-6-12-10(16)9(15)11(2,3)7-13/h9,13,15H,4-7H2,1-3H3,(H,12,16)/t9-/m0/s1
SMILES CCOC(=O)CCNC(=O)[C@H](O)C(C)(C)CO
IUPAC Name ethyl 3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate
Molecular Weight (g/mol) 247.29
Quantity 50 mg
Formula Weight 247.142 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.