missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Octanoate-d15, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl octanoate-d15 |
| CAS | 1219798-38-5 |
| Molecular Formula | C10 2H15 H5 O2 |
| Quantity | 50 mg |
| Synonym | Octanoic Acid-Ethyl Ester-D15 (octanoic-D15), Octanoic acid-ethyl ester-d15, Octanoic-d15 acid ethyl ester, Caprylic acid-d15 ethyl ester, Ethyl caprylate-d15, Ethyl octanoate-d15 |
| InChI Formula | InChI=1S/C10H20O2/c1-3-5-6-7-8-9-10(11)12-4-2/h3-9H2,1-2H3/i1D3,3D2,5D2,6D2,7D2,8D2,9D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OCC |
| IUPAC Name | ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoate |
| Molecular Weight (g/mol) | 187.36 |
| Formula Weight | 187.2405 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?