missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (E/Z)-3-(2-Carbethoxy-2-cyanoethenyl)amino-1H-pyrazole-4-carboxylate, Mikromol™
Description
Ethyl (E/Z)-3-(2-Carbethoxy-2-cyanoethenyl)amino-1H-pyrazole-4-carboxylate
Specifications
Specifications
| Chemical Name or Material | Ethyl (E/Z)-3-(2-Carbethoxy-2-cyanoethenyl)amino-1H-pyrazole-4-carboxylate |
| CAS | 321571-07-7 |
| Analyte or Component Names | Ethyl (E/Z)-3-(2-Carbethoxy-2-cyanoethenyl)amino-1H-pyrazole-4-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H14 N4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Pyrazole-4-carboxylic acid, 3-[(2-cyano-3-ethoxy-3-oxo-1-propen-1-yl)amino]-, ethyl ester, 1H-Pyrazole-4-carboxylic acid, 3-[(2-cyano-3-ethoxy-3-oxo-1-propenyl)amino]-, ethyl ester (9CI), (E/Z)-3-[(2-Cyano-3-ethoxy-3-oxo-1-propenyl)amino]-1H-Pyrazole-4-carboxylic Acid Ethyl Ester, Allopurinol Related Compound F (USP), Allopurinol Related Compound F |
| SMILES | CCOC(=O)C(=CNc1[nH]ncc1C(=O)OCC)C#N |
| Molecular Weight (g/mol) | 278.27 |
| Quantity | 25 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?