missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-nitrobenzoate, 99%

Product Code. 10154420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10154420 500 g Glass bottle 500g
10417010 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10154420 Supplier Thermo Scientific Acros Supplier No. 168425000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-77-4 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-77-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00007351
InChI Key PHWSCBWNPZDYRI-UHFFFAOYSA-N
Synonym ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate
PubChem CID 7457
IUPAC Name ethyl 4-nitrobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 56.0°C to 59.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CO2C2H5
Quantity 500 g
Beilstein 09, 390
Merck Index 10, 3776
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 195.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-nitrobenzoate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.