missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estradiol Cypionate, Mikromol™
Description
Estradiol Cypionate
Specifications
Specifications
| Chemical Name or Material | Estradiol Cypionate |
| CAS | 313-06-4 |
| Analyte or Component Names | Estradiol Cypionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H36 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Estradiol, 17-cyclopentanepropionate (6CI,7CI,8CI), Cyclopentanepropionic acid, 17-ester with estradiol, Depgynogen, Depo-Estradiol, Depo-estradiol cyclopentylpropionate, Depoestradiol, Depoestradiol cypionate, Depofemin, ECP, Estradep, Estradiol 17-cyclopentylpropionate, Estradiol 17-cypionate, Estradiol cyclopentylpropionate, Estradiol cypionate, Estradiolum valerianicum, NSC 3354, Cyclopentanepropionic acid, 3-hydroxyestra-1,3,5(10)-trien-17beta-yl ester (8CI), 17beta-Estradiol 17-cyclopentylpropionate, Estradiol 17beta-cyclopentanepropionate, Estradiol 17beta-cyclopentylpropionate, Estradiol 17beta-cypionate, Estra-1,3,5(10)-triene-3,17-diol (17beta)-, 17-cyclopentanepropanoate |
| InChI Formula | InChI=1S/C26H36O3/c1-26-15-14-21-20-10-8-19(27)16-18(20)7-9-22(21)23(26)11-12-24(26)29-25(28)13-6-17-4-2-3-5-17/h8,10,16-17,21-24,27H,2-7,9,11-15H2,1H3/t21-,22-,23+,24+,26+/m1/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5 |
| IUPAC Name | [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?