missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ESRRA Monoclonal Antibody (H5844)
GREENER_CHOICE

Product Code. 10125023
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10125023 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10125023 Supplier Invitrogen™ Supplier No. 416600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

This antibody specifically recognizes human ERR alpha and cross reacts with mouse and rat ERR alpha. This antibody does not recognize human ERR beta and gamma.

Estrogen-related receptor alpha 1 (ERR alpha, ERR1) is an orphan member of the nuclear hormone receptor superfamily. All the ERR family members share an almost identical DNA-binding domain, which has 68% amino acid identity with that of estrogen receptor. This protein binds to an ERR-alpha response element (ERRE) containing a single consensus half-site, 5'-tnaaggtca-3' can bind to the medium-chain acyl coenzyme a dehydrogenase (mcad) response element nrre-1 and may act as an important regulator of mcad promoter. Studies have shown that ERR1 functions a metabolic regulator and is often expressed in tissues that preferentially metabolize fatty acids. The corresponding gene for the ERR1 is NR3B1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRRA
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone H5844
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation physiological saline with 0.1% sodium azide
Gene ESRRA
Gene Accession No. O08580, P11474, Q5QJV7
Gene Alias ERR alpha; ERR1; ERRa; ERRalpha; ERR-alpha; Errra; ESRL1; ESRRA; ESRRA1; ESRRA2; estrogen receptor related 1; estrogen receptor-like 1; estrogen related receptor alpha; estrogen related receptor alpha 1; estrogen related receptor, alpha; estrogen-related nuclear receptor alpha; estrogen-related receptor alpha; estrogen-related receptor, alpha; Estrra; HERR1; LOW QUALITY PROTEIN: steroid hormone receptor ERR1; NR3B1; Nuclear receptor subfamily 3 group B member 1; orphan nuclear receptor; steroid hormone receptor ERR1; Steroid hormone receptor ERR1 (Estrogen-related receptor, alpha) (ERR-alpha) (Estrogen receptor-like 1)
Gene Symbols ESRRA
Host Species Mouse
Immunogen Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with Baculovirus-expressed recombinant human ERR--alpha 1 (98-171 aa).
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2101, 26379, 293701
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.