Learn More
Fisherbrand™ Erlenmeyer Flask Clamp for Fisherbrand™ Shaking Incubators
Product Code. 17749199
Change view
Click to view available options
Holds:
3 L Erlenmeyer Flask
Unit Size:
Each
Product Code. Holds unitSize
17749199 3 L Erlenmeyer Flask Each
1 options
Product Code. 17749199
Only null left
Add to basket
Add to basket
Erlenmeyer Flask Clamp for use with Fisherbrand™ Shaking Incubators with magnetic platform.
  • Magnetic attachment
  • Toolless exchange
  • Holds flask
Specifications
Holds 3 L Erlenmeyer Flask
For Use With (Equipment) Fisherbrand Shaking Incubator