Learn More
Sartorius Ergomate Pipet Accessories
Accessory for Ergomate Pipet
Brand: Sartorius 783500
58.14 GBP valid until 2024-12-31
Use promo code "23285" to get your promotional price.
Code : 04
Additional Details : Weight : 0.50000kg
Description
Ergomate removes the need to hold the plate at any angle and allows safe dispensing, resulting easier, safer and more precise pipetting.
According to ISO 8655 recommendations, dispensing especially small volumes should be done against the inside wall of receiving vessel. Ergomate is an ideal tool for finding proper positioning in pipetting.
Microplate grids for quick identification and sample management.
Grids are double sided. Reverse side featuring numbering from 1-12.
Plate Holder
When reversed plate holder creates an ideal aerosol barrier for use in hoods or on bench top surfaces.
Plate holder can be reversed and used for flat surface pipetting or as an incubation station with lid.
Ideal for safe transporting of plates.
Non autoclavable
Specifications
Autoclavable | |
120mL reagent vessel | |
All multichannel pipette |
Polypropylene | |
120 mL | |
120mL |