missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EPR1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10088479
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10088479 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10088479 Supplier Invitrogen™ Supplier No. PA513117

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPR1 Polyclonal Antibody for Western Blot

Cellular receptors for blood proteases regulate chemotaxis, extracellular proteolysis, and growth behavior of normal and malignant cells. Effector cell protease receptor-1 (EPR1) is a receptor for the coagulation protease factor Xa. EPR1 is characterized by a cysteine-rich extracellular module, a single membrane-spanning domain, and a serine-rich cytoplasmic tail featuring at least 15 potential phosphorylation sites. EPR1 also contains 2 N-linked glycosylation sites, 4 O-linked glycosylation sites, and a chondroitin sulfate attachment site, which may provide anchoring for carbohydrate chains, EPR1 transfectants bind to factor Xa in a specific and saturable manner, and in the absence of factor V/Va promote prothrombin activation in a factor Xa concentration-dependent reaction. Activated platelets and megakaryocytes express EPR1. Both EPR1 and membrane-bound factor Va are thought to be required to mediate factor Xa binding to the activated platelet to form a functional prothrombinase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias EPR-1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 308-337 amino acids from the C-terminal region of human EPR1.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.