missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EPHB1, GST Tag Recombinant Protein

Product Code. 10108252
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10108252 1 mg 1mg
15428473 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10108252 Supplier Invitrogen™ Supplier No. PV4100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic

EPHB1 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. ephB family proteins bind ephrins of the B class, ligands that are also transmembrane spanning proteins capable of transmitting signals. ephB1 is expressed predominantly in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, ephB1 signals regulation of cell attachment and cellcell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P54762
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 2047
Molecular Weight (g/mol) 60.3 kDa
Name Human EPHB1, GST Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias 9330129L11; AW488255; C130099E04Rik; Cek6; EK6; ELK; Elkh; ELK-related protein tyrosine kinase; ENSMUSG00000074119; Eph receptor B1; Eph receptor B2 (ELK-related protein tyrosine kinase); eph tyrosine kinase 2; EPHB1; Ephb2; EPH-like kinase 6; Ephrin type-B receptor 1; EPHT2; Epth2; Erk; Hek6; NET; Neuronally-expressed EPH-related tyrosine kinase; soluble EPHB1 variant 1; tyrosine-protein kinase receptor EPH-2
Common Name EphB1
KinaseFamily EPH Receptor Kinase Family
Protein Length 613-881
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.