missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EO 1428
GREENER_CHOICE

Product Code. 18065962
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065962 50 mg 50mg
18014602 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065962 Supplier Tocris Bioscience™ Supplier No. 2908/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of p38α and p38β2

Selective inhibitor of p38α and p38β2 that displays no activity at p38γ, p38δ, ERK1/2 and JNK1. Inhibits production of inflammatory cytokines including IL-8, TNF-α, IL-6, IL-1β and IL-10 (IC50 values are 4, 5, 17, 30 and 74 nM respectively). Displays anti-inflammatory activity in murine models of acute and chronic dermatitis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321351-00-2
Molecular Formula C20H16BrClN2O

Specifications

Quantity 50 mg
Formula Weight 415.71
Purity >98%
Molecular Weight (g/mol) 415.715
Chemical Name or Material EO 1428
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.