missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EO 1428

Description
Selective inhibitor of p38α and p38β2 that displays no activity at p38γ, p38δ, ERK1/2 and JNK1. Inhibits production of inflammatory cytokines including IL-8, TNF-α, IL-6, IL-1β and IL-10 (IC50 values are 4, 5, 17, 30 and 74 nM respectively). Displays anti-inflammatory activity in murine models of acute and chronic dermatitis.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 415.71 |
| Purity | >98% |
| Molecular Weight (g/mol) | 415.715 |
| Chemical Name or Material | EO 1428 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?