missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ENT2 Recombinant Rabbit Monoclonal Antibody (K01_1Q82)
GREENER_CHOICE

Product Code. 30829194
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829194 100 μL
30825921 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829194 Supplier Invitrogen™ Supplier No. MA563385

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ENT2 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

SLC29A2 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A2 is a transmembrane glycoprotein that mediates the cellular uptake of nucleosides from the surrounding medium. As a nucleoside transporter, SLC29A2 plays an important role in the uptake of nucleoside-based anti-cancer drugs; polymorphisms of point mutations in the gene encoding this protein may affect the efficacy of these drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENT2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1Q82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene SLC29A2
Gene Accession No. Q14542
Gene Alias 36 kDa hydrophobic nucleolar protein; 36 kDa nucleolar protein HNP36; delayed early response gene 12; delayed-early response protein 12; DER12; ENT2; equilibrative NBMPR-insensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter; equilibrative nucleoside transporter 2; HNP36; hydrophobic nucleolar protein, 36 kDa; hydrophobic nucleolar protein, 36kD; NBMPR-insensitive nucleoside transporter ei; NBMPR-insensitive nucleoside transporter ei 2A; nucleoside transporter, ei-type; rbENT2A; Slc29a2; solute carrier family 29 (equilibrative nucleoside transporter), member 2; solute carrier family 29 (nucleoside transporters), member 2; solute carrier family 29 member 2; solute carrier family 29, member 2
Gene Symbols SLC29A2
Host Species Rabbit
Immunogen A synthetic peptide of human ENT2.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3177
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.