Learn More
Merck Adapter, 1/8 NPTM to 1/4
Product Code. 10082133 Shop All Merck Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10082133 Water Purification System Each
1 options
Product Code. 10082133 Supplier Merck Supplier No. YT3009018
Only null left
Add to basket
Add to basket
Accessories for filter holders for sample preparation.
Adapts 1/8 or 1/4 in. NPTM fittings to 1/4 in. OD tubing
TRUSTED_SUSTAINABILITY
Specifications
Connection Type 1/4 in. tube O.D. to 1/8 in. NPTM
For Use With (Equipment) Water Purification System
Type Environmental Test Kits or Supplies
Material 316 stainless steel
Diameter (English) Inside 0.125 in.
Diameter (Metric) Inside 0.3175 cm