missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eicosanoic-17,17,18,18-d4 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Eicosanoic Acid-17,17,18,18-D4 |
| CAS | 1219803-69-6 |
| Molecular Formula | C20 D4 H36 O2 |
| Quantity | 0.1 g |
| Synonym | Eicosanoic-17,17,18,18-d4 acid, Eicosanoic Acid-17,17,18,18-D4, Eicosanoic acid-d4, Arachidic-17,17,18,18-d4 acid, Arachidic acid-17,17,18,18-d4, Arachidic acid-d4 |
| InChI Formula | InChI=1 S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-19H2,1H3,(H,21,22)/i3D2,4D2 |
| SMILES | [2 H]C([2 H])(CC)C([2 H])([2 H])CCCCCCCCCCCCCCCC(=O)O |
| IUPAC Name | 17,17,18,18-tetradeuterioicosanoic acid |
| Molecular Weight (g/mol) | 316.5551 |
| Formula Weight | 316.3279 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?