missing translation for 'onlineSavingsMsg'
Learn More

Edetate Disodium, U.S.P., J.T. Baker™

Product Code. 10199472 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10199472 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10199472 Supplier Avantor J.T.Baker Supplier No. 899401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Especificaciones

CAS 6381-92-6
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C10H18N2Na2O10
MDL Number MFCD00150037,MFCD00003541
Quantity 500 g
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Molecular Weight (g/mol) 372.24
PubChem CID 44120005
Formula Weight 372.24
Grade USP
Chemical Name or Material Edetate Disodium
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • EDTA disodium salt dihydrate U.S.P.
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Indicaciones de peligro
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P314-Get medical advice/attention if you feel unwell.
Otros peligros
  • MIXTURE LIST-Contain: EDTA disodium salt dihydrate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.