missing translation for 'onlineSavingsMsg'
Learn More

Edasalonexent, MedChemExpress

Product Code. 16499907
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499907 5 mg 5mg
16467406 100 mg 100mg
16463457 50 mg 50mg
16417438 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499907 Supplier Medchem Express Supplier No. HY17630

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Edasalonexent (CAT-1004) is an orally bioavailable NF-κB inhibitor.

Edasalonexent (CAT-1004) is an orally bioavailable NF-κB inhibitor.

Specifications

Chemical Name or Material Edasalonexent
CAS 1204317-86-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C31H42N2O3
Quantity 5 mg
Synonym CAT-1004
Solubility Information DMSO : 120 mg/mL (244.56 mM; Need ultrasonic)
SMILES O=C(NCCNC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)C1=CC=CC=C1O
Molecular Weight (g/mol) 490.68
Formula Weight 490.68
Percent Purity 98.0%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.