missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Eclipse™ 200 μL Clear Pipette Tips
GREENER_CHOICE

Product Code. 16651913
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Natural
Format:
Bagged
Rack, 192
Rack, 96
Refill
Stack Rack
Unit Size:
Pack of 1000
Pack of 200
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Color Format unitSize
16651913 Sterile Natural Bagged Pack of 200
16661913 Non-sterile Natural Bagged Pack of 1000
16603952 Non-sterile Clear Refill Pack of 960
16613952 Sterile Clear Rack, 96 Pack of 960
16671913 Sterile Natural Rack, 96 Pack of 960
17517826 Non-sterile Clear Stack Rack Pack of 960
17527826 Non-sterile Clear Rack, 192 Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16651913 Supplier Labcon™ Supplier No. 10601650009
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These beveled point pipette tips are available in racks, stack racks, refills, or bags.

  • Fits most standard 200 μL pipettes
  • Polypropylene USP Class VI Resin
  • Made in USA

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Length (English) 1.95 in.
Length (Metric) 49.7 mm
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims ATP, RNase/DNase, and Endotoxin-free
For Use With (Application) Liquid Handling
Sterilization Method E-beam
Format Bagged
Tip Style Beveled
Quantity 1 Tip/Bag, 200 Bags/Pk., 10 Pk./Cs.
Shipping Condition RT
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line Eclipse
Material Polypropylene Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.