missing translation for 'onlineSavingsMsg'
Learn More

Elma™ EC 75 Cleaner

Product Code. 10035361
Change view
Click to view available options
Volume (Metric):
25 L
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10035361 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10035361 Supplier Elma™ Supplier No. 5810610000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Providing powerful cleaning power for workshop and laboratory as well as for jewellery cleaning.

  • It cleans and brightens precious metals, copper, brass and gems, gives new brilliance.
  • Reliable removes dirt, fat, grease, sweat, oxides, buffing compound, rouge, lapping residues and polishing paste.
  • Lights up jewellery and non-ferrous metals due to ammonia.
  • Zinc, aluminum and light metal alloys may be attacked.
  • Not suitable for soft gems like pearls, corals, lapislazuli, amber, opal etc.

Specifications

Product Type EC 75 Cleaner
Certifications/Compliance CE, Annex VII, A, EC 648/2004
For Use With (Application) For cleaning jewellery, laboratory, workshops applications
Volume (Metric) 25 L
compliance-icons compliance-icons
Product Identifier
  • ELMACLEAN 75 CLEANING SOLUTION 25L .
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-methoxy-2-propanol, Fatty acids, C14-18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.