missing translation for 'onlineSavingsMsg'
Learn More

(E/Z)-3-[(2-Cyano-3-ethoxy-3-oxo-1-propenyl)amino]-1H-Pyrazole-4-carboxylic Acid Ethyl Ester, TRC

Product Code. 30349648
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349648 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349648 Supplier Toronto Research Chemicals Supplier No. TRCC979505100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (E/Z)-3-(2-Carbethoxy-2-cyanoethenyl)amino-1H-pyrazole-4-carboxylate
CAS 321571-07-7
Molecular Formula C12 H14 N4 O4
Quantity 100 mg
Synonym 1H-Pyrazole-4-carboxylic acid, 3-[(2-cyano-3-ethoxy-3-oxo-1-propen-1-yl)amino]-, ethyl ester, 1H-Pyrazole-4-carboxylic acid, 3-[(2-cyano-3-ethoxy-3-oxo-1-propenyl)amino]-, ethyl ester (9CI), (E/Z)-3-[(2-Cyano-3-ethoxy-3-oxo-1-propenyl)amino]-1H-Pyrazole-4-carboxylic Acid Ethyl Ester, Allopurinol Related Compound F (USP)
InChI Formula InChI=1S/C12H14N4O4/c1-3-19-11(17)8(5-13)6-14-10-9(7-15-16-10)12(18)20-4-2/h6-7H,3-4H2,1-2H3,(H2,14,15,16)
SMILES CCOC(=O)C(=CNc1[nH]ncc1C(=O)OCC)C#N
IUPAC Name ethyl 5-[(2-cyano-3-ethoxy-3-oxoprop-1-enyl)amino]-1H-pyrazole-4-carboxylate
Molecular Weight (g/mol) 278.264
Formula Weight 278.1015
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.