Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Product Code. 10002944
Change view
Click to view available options
Cone Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/31
NS 29/32
Standard Taper Joint No:
GL 14
GL 18
GL 25
GL 32
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Standard Taper Joint No unitSize
10002944 NS 29/32 GL 32 Each
10088450 NS 14/23 GL 14 Each
10597741 NS 19/26 GL 14 Each
10692411 NS 19/26 GL 18 Each
10324001 NS 19/26 GL 25 Each
10363901 NS 29/31 GL 14 Each
10708931 NS 24/29 GL 14 Each
10160121 NS 29/32 GL 25 Each
10405801 NS 24/29 GL 18 Each
10632421 NS 29/32 GL 18 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10002944 Supplier Lenz Laborglasinstrumente™ Supplier No. 01313229
Only null left
Add to basket
Add to basket
Provides for secure and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12257
  • Available in a range of socket and thread sizes
  • Minimized need for grease
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Quantity 1 Ea.
Cone Size NS 29/32
Standard Taper Joint No GL 32
Material Duran Glass