Learn More
Hecht Karl™ Duran™ Coil Condenser
Product Code. 10135761
Change view
Click to view available options
Standard Taper Joint No:
14/23
29/32
Length (Metric):
16cm
25cm
40cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) unitSize
10135761 29/32 40cm Each
10105381 14/23 16cm Each
10135521 29/32 25cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10135761 Supplier Hecht Karl™ Supplier No. 41996056